2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole
Supplier: Apollo Scientific
OR46716-1G
OR46716-250MG
OR46716-500MG
OR46716-2.5G
OR46716-5G
APOSOR46716-1GEA
439
EUR
APOSOR46716-1G
APOSOR46716-250MG
APOSOR46716-500MG
APOSOR46716-2.5G
APOSOR46716-5G
2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole
2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole
Formula:
C₆H₅N₃S Storage Temperature: Ambient |
MDL Number:
MFCD06653492 CAS Number: 881041-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...