3-Phenyl-1H-1,2,4-triazole

Supplier: Apollo Scientific

OR46717-1G OR46717-250MG OR46717-2.5G
APOSOR46717-1GEA 445 EUR
APOSOR46717-1G APOSOR46717-250MG APOSOR46717-2.5G
3-Phenyl-1H-1,2,4-triazole
3-Phenyl-1H-1,2,4-triazole
Formula: C₈H₇N₃
MW: 145,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159717
CAS Number: 3357-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR