4'-Acetamidoacetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Acetamidoacetophenone, N-(4-Acetylphenyl)acetamide
OR4673-1G
OR4673-25G
OR4673-5G
APOSOR4673-1GEA
21.9
EUR
APOSOR4673-1G
APOSOR4673-25G
APOSOR4673-5G
4'-Acetamidoacetophenone 98%
4'-Acetamidoacetophenone
Formula:
C₁₀H₁₁NO₂ MW: 177.20 g/mol Melting Pt: 166…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014965 CAS Number: 2719-21-3 EINECS: 220-320-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...