4-Iodoanisole 98%

Supplier: Apollo Scientific
Warning

OR4677-100G OR4677-25G
APOSOR4677-100GEA 87.3 EUR
APOSOR4677-100G APOSOR4677-25G
4-Iodoanisole 98%
4-Iodoanisole
Formula: C₇H₇IO
MW: 234,04 g/mol
Boiling Pt: 240 °C
Melting Pt: 48…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001056
CAS Number: 696-62-8
EINECS: 211-798-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR