BippyPhos 97%
Supplier: Apollo Scientific
Synonyms:
5-(DI-T-BUTYLPHOSPHINO)-1',3',5'-TRIPHENYL-1,4'-BI-1H-PYRAZOLE, 5-(Di-tert-butylphosphino)-1', 3', 5'-triphenyl-1'H-[1,4']bipyrazole, 5-(di-tert-Butylphosphino)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole, 5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole
OR46775-1G
OR46775-5G
APOSOR46775-1GEA
109
EUR
APOSOR46775-1G
APOSOR46775-5G
BippyPhos 97%
BippyPhos
Formula:
C₃₂H₃₅N₄P MW: 506.634 g/mol Melting Pt: 169…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD09038440 CAS Number: 894086-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...