2-Iodoanisole 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Iodo-2-methoxybenzene
OR4678-500G
APOSOR4678-500GEA
231
EUR
APOSOR4678-500G
2-Iodoanisole 98%
2-Iodoanisole
Formula:
C₇H₇IO MW: 234,04 g/mol Boiling Pt: 240…242 °C Density: 1,799 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001039 CAS Number: 529-28-2 EINECS: 208-456-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...