1-Iodundecane

Supplier: Apollo Scientific
Warning

OR4680-10G OR4680-5G
APOSOR4680-10GEA 143 EUR
APOSOR4680-10G APOSOR4680-5G
1-Iodundecane
1-Iodundecane
Formula: C₁₁H₂₃I
MW: 282.21 g/mol
Boiling Pt: 125…130 °C/5mmHg
Melting Pt: –15.99 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
CAS Number: 4282-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR