3-(Boc-aminomethyl)pyridine

Supplier: Apollo Scientific

Synonyms: tert-butyl (pyridin-3-ylmethyl)carbamate

OR470015-25G OR470015-5G
APOSOR470015-25GEA 448 EUR
APOSOR470015-25G APOSOR470015-5G
3-(Boc-aminomethyl)pyridine
3-(Boc-aminomethyl)pyridine
Formula: C₁₁H₁₆N₂O₂
MW: 208,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411296
CAS Number: 102297-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR