3-(2-Nitrophenoxy)propionic acid

Supplier: Apollo Scientific

OR470097-1G OR470097-5G
APOSOR470097-1GEA 256 EUR
APOSOR470097-1G APOSOR470097-5G
3-(2-Nitrophenoxy)propionic acid
3-(2-Nitrophenoxy)propionic acid
Formula: C₉H₉NO₅
Storage Temperature: Ambient
MDL Number: MFCD09045698
CAS Number: 6336-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR