(4-Methylphenoxy)acetonitrile
Supplier: Apollo Scientific
|
Warning
|
OR470168-1G
OR470168-5G
APOSOR470168-1GEA
40.8
EUR
APOSOR470168-1G
APOSOR470168-5G
(4-Methylphenoxy)acetonitrile
(4-Methylphenoxy)acetonitrile
Formula:
C₉H₉NO MW: 147,18 g/mol Boiling Pt: 73…78 °C/11mmHg Melting Pt: 42…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068121 CAS Number: 33901-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...