5-Amino-3-(3,5-dichlorophenyl)isoxazole
Supplier: Apollo Scientific
OR470199-1G
OR470199-5G
APOSOR470199-1GEA
306
EUR
APOSOR470199-1G
APOSOR470199-5G
5-Amino-3-(3,5-dichlorophenyl)isoxazole
5-Amino-3-(3,5-dichlorophenyl)isoxazole
Formula:
C₉H₆Cl₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD11156303 CAS Number: 1020997-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...