1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
Supplier: Apollo Scientific
OR470210-25G
OR470210-5G
APOSOR470210-25GEA
1540
EUR
APOSOR470210-25G
APOSOR470210-5G
1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
Formula:
C₉H₁₆N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD03830326 CAS Number: 702670-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...