2',3',4'-Trimethoxyacetophenone 97%

Supplier: Apollo Scientific

Synonyms: 1-(2,3,4-Trimethoxyphenyl)ethan-1-one, 2-3-4-Trimethxoyacetophenone

OR4705-5G OR4705-25G
APOSOR4705-5GEA 101 EUR
APOSOR4705-5G APOSOR4705-25G
2',3',4'-Trimethoxyacetophenone 97%
2',3',4'-Trimethoxyacetophenone
Formula: C₁₁H₁₄O₄
MW: 210,23 g/mol
Boiling Pt: 295…297 °C
Melting Pt: 14…15 °C
Density: 1,155 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008726
CAS Number: 13909-73-4
EINECS: 237-678-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR