2,4,6-Triiodophenol
Supplier: Apollo Scientific
OR4709-25G
OR4709-5G
OR4709-100G
APOSOR4709-25GEA
125
EUR
APOSOR4709-25G
APOSOR4709-5G
APOSOR4709-100G
2,4,6-Triiodophenol
2,4,6-Triiodophenol
Formula:
C₆H₃I₃O MW: 471,8 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002179 CAS Number: 609-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...