2-Amino-4-cyanopyrimidine

Supplier: Apollo Scientific

Synonyms: 2-Aminopyrimidine-4-carbonitrile

OR471597-1G OR471597-5G
APOSOR471597-1GEA 118 EUR
APOSOR471597-1G APOSOR471597-5G
2-Amino-4-cyanopyrimidine
2-Amino-4-cyanopyrimidine
Formula: C₅H₄N₄
Storage Temperature: Ambient
MDL Number: MFCD09834971
CAS Number: 36314-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR