D-α-Aminoadipic acid 97%
Supplier: Apollo Scientific
Synonyms:
D-2-Aminoadipic acid, (R)-2-Aminohexanedioic acid, H-D-Aad-OH
OR471645-25G
APOSOR471645-25GEA
63.8
EUR
APOSOR471645-25G
D-α-Aminoadipic acid 97%
D-α-Aminoadipic acid
Formula:
C₆H₁₁NO₄ MW: 161,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063118 CAS Number: 7620-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...