6-(Chloromethyl)pyridine-2-carbonitrile 95%

Supplier: Apollo Scientific

Synonyms: 6-(Chloromethyl)-2-cyanopyridine

OR471675-5G
APOSOR471675-5GEA 140 EUR
APOSOR471675-5G
6-(Chloromethyl)pyridine-2-carbonitrile 95%
6-(Chloromethyl)pyridine-2-carbonitrile
Formula: C₇H₅ClN₂
MW: 152,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991809
CAS Number: 135450-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR