2-Bromotoluene 99%
Supplier: Apollo Scientific
|
Warning
|
OR4737-100G
OR4737-500G
APOSOR4737-100GEA
52.2
EUR
APOSOR4737-100G
APOSOR4737-500G
2-Bromotoluene 99%
2-Bromotoluene
Formula:
C₇H₇Br Boiling Pt: 181 °C/760mmHg Melting Pt: –27 °C Density: 1,422 g/cm³ (25 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000068 CAS Number: 95-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...