2-(Dimethoxymethyl)-1,6-naphthyridine 95+%

Supplier: Apollo Scientific
Warning

OR4773-1G OR4773-250MG
APOSOR4773-1GEA 140 EUR
APOSOR4773-1G APOSOR4773-250MG
2-(Dimethoxymethyl)-1,6-naphthyridine 95+%
2-(Dimethoxymethyl)-1,6-naphthyridine
Formula: C₁₁H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00202920
CAS Number: 386715-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR