2-Amino-4-bromopyridine 95%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-pyridylamine, 4-Bromopyridin-2-amine

OR4774-1G OR4774-5G
APOSOR4774-1GEA 21.9 EUR
APOSOR4774-1G APOSOR4774-5G
2-Amino-4-bromopyridine 95%
2-Amino-4-bromopyridine
Formula: C₅H₅BrN₂
MW: 173,01 g/mol
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD01646115
CAS Number: 84249-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR