Benzyl chloromethyl ether

Supplier: Apollo Scientific

Synonyms: ((Chloromethoxy)methyl)benzene

OR47887-100G OR47887-1KG OR47887-250G
APOSOR47887-100GEA 256 EUR
APOSOR47887-100G APOSOR47887-1KG APOSOR47887-250G
Benzyl chloromethyl ether
Benzyl chloromethyl ether
Formula: C₈H₉ClO
MW: 156,61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00000886
CAS Number: 3587-60-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR