DL-4-Methoxymandelic acid
Supplier: Apollo Scientific
|
Warning
|
OR4792-25G
OR4792-5G
APOSOR4792-25GEA
46.4
EUR
APOSOR4792-25G
APOSOR4792-5G
DL-4-Methoxymandelic acid
DL-4-Methoxymandelic acid
Formula:
C₉H₁₀O₄ Melting Pt: 108…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004233 CAS Number: 10502-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...