2-Chloro-5-phenylpyrazine 95%

Supplier: Apollo Scientific

OR480225-1G OR480225-500MG OR480225-5G
APOSOR480225-1GEA 292 EUR
APOSOR480225-1G APOSOR480225-500MG APOSOR480225-5G
2-Chloro-5-phenylpyrazine 95%
2-Chloro-5-phenylpyrazine
Formula: C₁₀H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD03268585
CAS Number: 25844-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR