2-(4-Methoxyphenyl)decahydroquinoxaline 95%
Supplier: Apollo Scientific
Synonyms:
2-(4-Methoxyphenyl)-decahydroquinoxaline, mixture cis/trans isomers
OR480293-1G
OR480293-250MG
OR480293-500MG
APOSOR480293-1GEA
350
EUR
APOSOR480293-1G
APOSOR480293-250MG
APOSOR480293-500MG
2-(4-Methoxyphenyl)decahydroquinoxaline 95%
2-(4-Methoxyphenyl)decahydroquinoxaline
Formula:
C₁₅H₂₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD07021286 CAS Number: 1005275-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...