4-(4-Hydroxyphenyl)cyclohexanone 97%

Supplier: Apollo Scientific
Warning

OR480590-100G OR480590-25G
APOSOR480590-100GEA 150 EUR
APOSOR480590-100G APOSOR480590-25G
4-(4-Hydroxyphenyl)cyclohexanone 97%
4-(4-Hydroxyphenyl)cyclohexanone
Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00210693
CAS Number: 105640-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR