Ethyl-5-methylisoxazole-4-carboxylate 97%
Supplier: Apollo Scientific
Synonyms:
Ethyl 5-methylisoxazole-4-carboxylate
OR480656-25G
OR480656-50G
APOSOR480656-25GEA
216
EUR
APOSOR480656-25G
APOSOR480656-50G
Ethyl-5-methylisoxazole-4-carboxylate 97%
Ethyl 5-methylisoxazole-4-carboxylate
Formula:
C₇H₉NO₃ MW: 155,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631119 CAS Number: 51135-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...