2-Cyclohexen-1-ol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Cyclohex-2-ene-1-ol, Cyclohex-2-enol
OR480699-25G
OR480699-5G
APOSOR480699-25GEA
330
EUR
APOSOR480699-25G
APOSOR480699-5G
2-Cyclohexen-1-ol 97%
2-Cyclohexen-1-ol
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 166 °C Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001570 CAS Number: 822-67-3 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...