2-Acetyl-5-nitrobenzofuran 98%

Supplier: Apollo Scientific

OR480704-25G OR480704-5G
APOSOR480704-25GEA 708 EUR
APOSOR480704-25G APOSOR480704-5G
2-Acetyl-5-nitrobenzofuran 98%
2-Acetyl-5-nitrobenzofuran
Formula: C₁₀H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00051634
CAS Number: 23136-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR