4-Methoxy-2-methylbenzaldehyde 97%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-4-methoxybenzaldehyde

OR480706-25G OR480706-5G
APOSOR480706-25GEA 224 EUR
APOSOR480706-25G APOSOR480706-5G
4-Methoxy-2-methylbenzaldehyde 97%
4-Methoxy-2-methylbenzaldehyde
Formula: C₉H₁₀O₂
MW: 150,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02261771
CAS Number: 52289-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR