2,6-Dichloropyrimidine-4-carboxylic acid 97%

Supplier: Apollo Scientific

OR480754-5G
APOSOR480754-5GEA 148 EUR
APOSOR480754-5G
2,6-Dichloropyrimidine-4-carboxylic acid 97%
2,6-Dichloropyrimidine-4-carboxylic acid
Formula: C₅H₂Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02933142
CAS Number: 16492-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR