6-Chloro-8-nitro-4H-benzo[1,4]oxazin-3-one 95%

Supplier: Apollo Scientific

OR480794-1G OR480794-5G
APOSOR480794-1GEA 115 EUR
APOSOR480794-1G APOSOR480794-5G
6-Chloro-8-nitro-4H-benzo[1,4]oxazin-3-one 95%
6-Chloro-8-nitro-4H-benzo[1,4]oxazin-3-one
Formula: C₈H₅ClN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD11226538
CAS Number: 870064-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR