Ethyl-1H-indazole-7-carboxylate 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl 1H-indazole-7-carboxylate
OR480801-1G
OR480801-5G
APOSOR480801-1GEA
64
EUR
APOSOR480801-1G
APOSOR480801-5G
Ethyl-1H-indazole-7-carboxylate 95%
Ethyl 1H-indazole-7-carboxylate
Formula:
C₁₀H₁₀N₂O₂ MW: 190,2000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07371560 CAS Number: 885278-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...