Methyl-2-amino-4-oxazolecarboxylate 95%

Supplier: Apollo Scientific

OR480826-1G OR480826-5G
APOSOR480826-1GEA 0 EUR
APOSOR480826-1G APOSOR480826-5G
Methyl-2-amino-4-oxazolecarboxylate 95%
Methyl 2-amino-4-oxazolecarboxylate
Formula: C₅H₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09870069
CAS Number: 1000576-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR