4-Bromo-2-(methylthio)pyrimidine 97%

Supplier: Apollo Scientific

Synonyms: 4-Bromo-2-methylsulphanyl-pyrimidine

OR480843-1G OR480843-5G
APOSOR480843-1GEA 121 EUR
APOSOR480843-1G APOSOR480843-5G
4-Bromo-2-(methylthio)pyrimidine 97%
4-Bromo-2-(methylthio)pyrimidine
Formula: C₅H₅BrN₂S
MW: 205,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878932
CAS Number: 959236-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR