4-Phenyl-1-butyne

Supplier: Apollo Scientific

OR48109-25G OR48109-5G
APOSOR48109-25GEA 554 EUR
APOSOR48109-25G APOSOR48109-5G
4-Phenyl-1-butyne
4-Phenyl-1-butyne
Formula: C₁₀H₁₀
MW: 130,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041667
CAS Number: 16520-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR