2'-Bromo-4',5'-(ethylenedioxy)acetophenone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)ethanone

OR4822-1G OR4822-250MG OR4822-5G
APOSOR4822-1GEA 205 EUR
APOSOR4822-1G APOSOR4822-250MG APOSOR4822-5G
2'-Bromo-4',5'-(ethylenedioxy)acetophenone 98%
2'-Bromo-4',5'-(ethylenedioxy)acetophenone
Formula: C₁₀H₉BrO₃
MW: 257.0807 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00168681
CAS Number: 59820-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR