N,N-Dimethylformamide diethyl acetal 90%

Supplier: Apollo Scientific
Warning

Synonyms: 1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine

OR48241-25G OR48241-100G
APOSOR48241-25GEA 85.8 EUR
APOSOR48241-25G APOSOR48241-100G
N,N-Dimethylformamide diethyl acetal 90%
N,N-Dimethylformamide diethyl acetal
Formula: C₇H₁₇NO₂
MW: 147,22 g/mol
Boiling Pt: 130…136 °C
Density: 0,859 g/cm³ (25 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00009227
CAS Number: 1188-33-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR