3-Nitro-1H-1,2,4-triazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Nitro-1,2,4-1H-triazole, 3-Nitro-1,2,4-triazole
OR4862-25G
OR4862-5G
APOSOR4862-25GEA
36.4
EUR
APOSOR4862-25G
APOSOR4862-5G
3-Nitro-1H-1,2,4-triazole
3-Nitro-1H-1,2,4-triazole
Formula:
C₂H₂N₄O₂ MW: 114,06 g/mol Melting Pt: 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00009749 CAS Number: 24807-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...