2-(Methylthio)ethylamine

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Methylthio)ethanamine

OR4873-1G OR4873-5G OR4873-25G
APOSOR4873-1GEA 26.1 EUR
APOSOR4873-1G APOSOR4873-5G APOSOR4873-25G
2-(Methylthio)ethylamine
2-(Methylthio)ethylamine
Formula: C₃H₉NS
Boiling Pt: 146…149 °C
Density: 0,974 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Freezer
MDL Number: MFCD00014825
CAS Number: 18542-42-2
EINECS: 242-412-5
UN: 2734
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR