4-Bromo-3-methylanisole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene
OR4933-10G
OR4933-25G
OR4933-5G
APOSOR4933-10GEA
21.9
EUR
APOSOR4933-10G
APOSOR4933-25G
APOSOR4933-5G
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole
Formula:
C₈H₉BrO MW: 201,06 g/mol Boiling Pt: 111…112 °C/15mmHg Density: 1,424 g/cm³ (25 °C) Flash Pt: >113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060687 CAS Number: 27060-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...