3',5'-Dibenzyloxyacetophenone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-[3,5-Bis(benzyloxy)phenyl]ethan-1-one

OR4950-10G OR4950-250G OR4950-50G
APOSOR4950-10GEA 71.3 EUR
APOSOR4950-10G APOSOR4950-250G APOSOR4950-50G
3',5'-Dibenzyloxyacetophenone 98%
3',5'-Dibenzyloxyacetophenone
Formula: C₂₂H₂₀O₃
MW: 332,40 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00004777
CAS Number: 28924-21-2
EINECS: 249-315-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR