4-tert-Butylaniline 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(tert-Butyl)aniline

OR4958-25G OR4958-5G OR4958-100G
APOSOR4958-25GEA 17.6 EUR
APOSOR4958-25G APOSOR4958-5G APOSOR4958-100G
4-tert-Butylaniline 98%
4-tert-Butylaniline
Formula: C₁₀H₁₅N
MW: 149,24 g/mol
Boiling Pt: 90…93 °C/3mmHg
Melting Pt: 15…16 °C
Density: 0,937 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007899
CAS Number: 769-92-6
EINECS: 212-215-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR