(2-Chlorophenyl)acetonitrile 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Chlorobenzyl cyanide, 2-(2-Chlorophenyl)acetonitrile
OR4975-100G
OR4975-25G
APOSOR4975-100GEA
49.5
EUR
APOSOR4975-100G
APOSOR4975-25G
(2-Chlorophenyl)acetonitrile 97%
(2-Chlorophenyl)acetonitrile
Formula:
C₈H₆ClN MW: 151,6 g/mol Boiling Pt: 240…242 °C Melting Pt: 21…24 °C Density: 1,17 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001898 CAS Number: 2856-63-5 EINECS: 220-669-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...