Piperonylic acid 98%

Supplier: Apollo Scientific
Danger

Synonyms: 3,4-(Methylenedioxy)benzoic acid, Benzo[d][1,3]dioxole-5-carboxylic acid, 1,3-Benzodioxole-5-carboxylic acid

OR4989-25G OR4989-100G
APOSOR4989-25GEA 56.8 EUR
APOSOR4989-25G APOSOR4989-100G
Piperonylic acid 98%
Piperonylic acid
Formula: C₈H₆O₄
MW: 166,13 g/mol
Melting Pt: 229…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00005830
CAS Number: 94-53-1
EINECS: 202-342-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR