5-Nitrosalicylic acid 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-5-nitrobenzoic acid
OR4992-100G
OR4992-25G
OR4992-500G
APOSOR4992-100GEA
102
EUR
APOSOR4992-100G
APOSOR4992-25G
APOSOR4992-500G
5-Nitrosalicylic acid 99%
5-Nitrosalicylic acid
Formula:
C₇H₅NO₅ MW: 183,12 g/mol Melting Pt: 228…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007338 CAS Number: 96-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...