8-Quinolineboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: Quinolin-8-ylboronic acid

OR4993-25G OR4993-5G OR4993-10G
APOSOR4993-25GEA 0 EUR
APOSOR4993-25G APOSOR4993-5G APOSOR4993-10G
8-Quinolineboronic acid 98%
8-Quinolineboronic acid

Exists as dimer.Note: NMR spectrum must be run in D2O.

Formula: C₉H₈BNO₂
MW: 172,98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01114698
CAS Number: 86-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR