4-Chlorobenzyl alcohol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Chlorophenyl)methanol
OR5022-25G
OR5022-500G
APOSOR5022-25GEA
21.9
EUR
APOSOR5022-25G
APOSOR5022-500G
4-Chlorobenzyl alcohol 98%
4-Chlorobenzyl alcohol
Formula:
C₇H₇ClO MW: 142,58 g/mol Boiling Pt: 234 °C Melting Pt: 68…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004652 CAS Number: 873-76-7 EINECS: 212-852-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...