3-Pyridineacetic acid 99%

Supplier: Apollo Scientific
Warning

Synonyms: (Pyridin-3-yl)acetic acid

OR5061-25G OR5061-5G OR5061-100G
APOSOR5061-25GEA 172 EUR
APOSOR5061-25G APOSOR5061-5G APOSOR5061-100G
3-Pyridineacetic acid 99%
3-Pyridineacetic acid
Formula: C₇H₇NO₂
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00066302
CAS Number: 501-81-5
EINECS: 207-928-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR