2,4,6-Trichloropyrimidine 98%

Supplier: Apollo Scientific
Danger

OR5063-100G OR5063-25G OR5063-5G
APOSOR5063-100GEA 53.8 EUR
APOSOR5063-100G APOSOR5063-25G APOSOR5063-5G
2,4,6-Trichloropyrimidine 98%
2,4,6-Trichloropyrimidine
Formula: C₄HCl₃N₂
MW: 183.42 g/mol
Boiling Pt: 210…215 °C
Melting Pt: 23…25 °C
Density: 1.595 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006063
CAS Number: 3764-01-0
EINECS: 223-183-0
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR