2-Chloro-6-methylnicotinic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR5068-25G
OR5068-5G
OR5068-100G
APOSOR5068-25GEA
33.5
EUR
APOSOR5068-25G
APOSOR5068-5G
APOSOR5068-100G
2-Chloro-6-methylnicotinic acid 98%
2-Chloro-6-methylnicotinic acid
Formula:
C₇H₆ClNO₂ MW: 171,58 g/mol Melting Pt: 157…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134165 CAS Number: 30529-70-5 EINECS: 250-229-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...